Order Entry
Hungary
ContactUsLinkComponent
EPCAM (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human EPCAM in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5064
Supplier:  Abnova
EPCAM (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human EPCAM in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5064
Supplier:  Abnova
Frequently Bought Together