Order Entry
Hungary
ContactUsLinkComponent
Ccl27 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse CCL27 in cell culture su pernates, cell lysates, tissue homogenates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5060
Supplier:  Abnova
Ccl27 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse CCL27 in cell culture su pernates, cell lysates, tissue homogenates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5060
Supplier:  Abnova
Frequently Bought Together