Order Entry
Hungary
ContactUsLinkComponent
Lif (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Lif in cell culture supern ates, cell lysates, tissue homogenates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5057
Supplier:  Abnova
Lif (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Lif in cell culture supern ates, cell lysates, tissue homogenates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5057
Supplier:  Abnova
Frequently Bought Together