Order Entry
Hungary
ContactUsLinkComponent
With the screw-on grinding bowl lid made of hardened, stainless steel, you can seal the hardened, stainless steel grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the h 1 * 1 items
Catalog # FRIT50.6555.00
Supplier:  FRITSCH
With the screw-on grinding bowl lid made of hardened, stainless steel, you can seal the hardened, stainless steel grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the h 1 * 1 items
Catalog # FRIT50.6555.00
Supplier:  FRITSCH
Frequently Bought Together