Order Entry
Hungary
ContactUsLinkComponent
Precision therm., low temp., similar DIN, -38+50:1C, solid stem, white backed, red special liquid, 220x6-7mm, with ring at top, sui table for government verification 1 * 1 items
Catalog # AMARL33088-FL
Gyártó:  Amarell
Precision therm., low temp., similar DIN, -38+50:1C, solid stem, white backed, red special liquid, 220x6-7mm, with ring at top, sui table for government verification 1 * 1 items
Catalog # AMARL33088-FL
Gyártó:  Amarell
Frequently Bought Together