Order Entry
Hungary
ContactUsLinkComponent
Thermometer similar to BS 593, F300C/76, -5+300:1C, solid stem, white backed, gallium filled, 390mm long, 76mm immersion, durable p igment, suitable for government verification 1 * 1 items
Catalog # AMARA403406-GA
Supplier:  Amarell
Thermometer similar to BS 593, F300C/76, -5+300:1C, solid stem, white backed, gallium filled, 390mm long, 76mm immersion, durable p igment, suitable for government verification 1 * 1 items
Catalog # AMARA403406-GA
Supplier:  Amarell
Frequently Bought Together