Order Entry
Hungary
Orders LinkContactUsLinkComponent
Human Recombinant Tissue Plasminogen Activator

Human Recombinant Tissue Plasminogen Activator

1 mg
Catalog # APOSBITP1356-1MG
Supplier:   Apollo Scientific
Human Recombinant Tissue Plasminogen Activator
Catalog # APOSBITP1356-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human Recombinant Tissue Plasminogen Activator

1 mg

CAS Number:  
Price
1 037 000 Ft
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1356-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Tissue Plasminogen Activator
  • Protein synonyms:
    tPA
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.