Order Entry
Hungary
ContactUsLinkComponent
PTFE-encapsulated magnetic core (Alnico 5), tapered blade with fixed cylindrical stirring bar, universal chemical resistance. Length x Width mm 8,0 x 8,0. Height mm 11, For vessel Int. Ø mm 9-10 1 * 3 items
Catalog # BOHLC349-04
Supplier:  Bohlender
undefined
PTFE-encapsulated magnetic core (Alnico 5), tapered blade with fixed cylindrical stirring bar, universal chemical resistance. Length x Width mm 8,0 x 8,0. Height mm 11, For vessel Int. Ø mm 9-10 1 * 3 items
Catalog # BOHLC349-04
Supplier:  Bohlender
Frequently Bought Together