Order Entry
Hungary
ContactUsLinkComponent
Sodium periodate ?99,8 %, p.a., ACS Sodium meta periodate Empirical formula NaIO4 Molar mass (M) 213,89 g/mol Density (D) 3,87 Melting point (mp)300 °C (dec.) Solubility 144 g/l (H2O, 25 °C) 1 * 250 g
Catalog # ROTH2603.2
Supplier:  Roth Carl
Sodium periodate ?99,8 %, p.a., ACS Sodium meta periodate Empirical formula NaIO4 Molar mass (M) 213,89 g/mol Density (D) 3,87 Melting point (mp)300 °C (dec.) Solubility 144 g/l (H2O, 25 °C) 1 * 250 g
Catalog # ROTH2603.2
Supplier:  Roth Carl
Frequently Bought Together