Order Entry
Hungary
ContactUsLinkComponent
Cogent HPS Phenyl HPLC Column 120A 5um 4.6mm x 250mm with Standard End Fittings. 1 * 1 items
Catalog # MICS75020-25P
Supplier:  MICROSOLV
undefined
Cogent HPS Phenyl HPLC Column 120A 5um 4.6mm x 250mm with Standard End Fittings. 1 * 1 items
Catalog # MICS75020-25P
Supplier:  MICROSOLV
Frequently Bought Together