Order Entry
Hungary
ContactUsLinkComponent
Cogent HPS Phenyl HPLC Column 120A 5um 2.1mm x 150mm with Standard End Fittings. 1 * 1 items
Catalog # MICS75020-15P-2
Supplier:  MICROSOLV
undefined
Cogent HPS Phenyl HPLC Column 120A 5um 2.1mm x 150mm with Standard End Fittings. 1 * 1 items
Catalog # MICS75020-15P-2
Supplier:  MICROSOLV
Frequently Bought Together