Cogent HPS C18 HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 100mm. 1 * 1 items
MICS75018-P100
Cogent HPS C18 HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 100mm. 1 * 1 items
MICS75018-P100
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection