Order Entry
Hungary
ContactUsLinkComponent
Easy Purchase Pack, 2ml clear vials with RSA-Pro glass treatment (12x32mm) with write on patch and light blue AQR snap caps with fit ted Silicone/PTFE septa. RSA-Pro Brand. 100/pack 1 * 100 items
Catalog # MICS9509C-1WDCP-MR
Supplier:  MICROSOLV
undefined
Easy Purchase Pack, 2ml clear vials with RSA-Pro glass treatment (12x32mm) with write on patch and light blue AQR snap caps with fit ted Silicone/PTFE septa. RSA-Pro Brand. 100/pack 1 * 100 items
Catalog # MICS9509C-1WDCP-MR
Supplier:  MICROSOLV
Frequently Bought Together