0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
MULTITEX SOFT ROLLE WHITE 1 * 2 items
ZVGA11801-01
:
Zellstoff-Vertriebs-GmbH & Co. KG
MULTITEX SOFT ROLLE WHITE 1 * 2 items
ZVGA11801-01
:
Zellstoff-Vertriebs-GmbH & Co. KG
Frequently Bought Together
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
: VWR Collection
pH electrodes, SenTix® PLUS
: WTW
Facial tissues, KLEENEX®
: KIMBERLY CLARK
VWR® pHenomenal® pH Electrodes
: VWR Collection
Electrolyte Solutions
: HAMILTON BONADUZ
pH electrodes, InLab®, Expert Series
: Mettler - Toledo
Kitchen rolls, Tork Premium
: Essity
pH electrodes, InLab® ISM
: Mettler - Toledo
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
: KIMBERLY CLARK
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
: VWR Collection
VWR®, Bench Protectors
: VWR Collection