Ugrás ide Fejléc
Ugrás ide Fő tartalom
Ugrás ide Lábléc
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Gyors rendelés
Megrendelések állapota
Saját fiók
Kosár
Hungary
Gyors rendelés
Megrendelések állapota
Saját fiók
Kosár
ContactUsLinkComponent
SPE-polypropylene columns C18 CHROMABOND® C18 ec Pore size: 60 Å, particle size: 45 µm, 3ml, 500mg sorbent 5x50/pck 250pcs/pck 1 * 250 items
Catalog # ROTHN748.2
Supplier:
Roth Carl
Expand image
SPE-polypropylene columns C18 CHROMABOND® C18 ec Pore size: 60 Å, particle size: 45 µm, 3ml, 500mg sorbent 5x50/pck 250pcs/pck 1 * 250 items
Catalog # ROTHN748.2
Supplier:
Roth Carl
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals