Order Entry
Hungary
Orders LinkContactUsLinkComponent
COD standards

COD standards

COD standard solution, 20 mg/l, 500 mL
Catalog # REGECOD20
Supplier:   REAGECON
COD standards
Catalog # REGECOD20
Supplier:  REAGECON
CAS Number:  
COD standards

COD standard solution, 20 mg/l, 500 mL

CAS Number:  
Price
61 700 Ft
Bejelentkezés az elérhetőségért
1 * 500 mL
Product Details & Documents

Ideal for use as control standards to verify that correct analysis for COD has taken place.

Documents

Specifikációk

Specifikációk

  • Concentration:
    20 mg/l
  • Pk:
    500 mL
  • Description:
    COD standard solution

Specifikációk

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.