Order Entry
Hungary
ContactUsLinkComponent
Pepsin?2000 FIP-U/g, for biochemistry From porcine gastric mucosa. 250g For detection of trichina in meat. Molar mass (M) 35000 g/mol Storage temp.: +4 °C WGK 1 CAS-Nr. [9001-75-6] EG-Nr. 232-629-3 1 * 250 g
Catalog # ROTHKK38.3
Supplier:  Roth Carl
Pepsin?2000 FIP-U/g, for biochemistry From porcine gastric mucosa. 250g For detection of trichina in meat. Molar mass (M) 35000 g/mol Storage temp.: +4 °C WGK 1 CAS-Nr. [9001-75-6] EG-Nr. 232-629-3 1 * 250 g
Catalog # ROTHKK38.3
Supplier:  Roth Carl
Frequently Bought Together