Boat-combustion Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), without handle, translucent, high chemic al resistance and purity. length 103mm, width 20mm, height 12mm. The durability or operating cycles of our Boat-combus 1 * 1 items
Catalog # TSLA73372
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together