Cryo Boxes Ratiolab® with Slip-Lid, Format 133 x 133 mm, polypropylene, yellow color. Adjustable in height from 50 mm to 75 mm, autoclavable, grid 7 x 7. For tube Ø 17 mm. 1 * 5 items
Katalógus# RATI5320311
Supplier: RATIOLAB
Frequently Bought Together