Order Entry
Hungary
ContactUsLinkComponent
GOAT AFFINITY PURIFIED ANTIBODY TO HUMAN IgG Fc. Anti-Human Host: goat Form: affinity purified liquid Applications: Immunostaining o f acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 1 mg
Catalog # ICNA0855071
Supplier:  MP Biomedicals
GOAT AFFINITY PURIFIED ANTIBODY TO HUMAN IgG Fc. Anti-Human Host: goat Form: affinity purified liquid Applications: Immunostaining o f acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 1 mg
Catalog # ICNA0855071
Supplier:  MP Biomedicals
Frequently Bought Together