Order Entry
Hungary
ContactUsLinkComponent
ETHIDIUM BROMIDE. Molecular Biology ReagentPurity: 98%Ideal for fluorometric detection of double stranded nucleic acids in gel elect rophoresis. Also acts as an RNA polymerase inhibitor, and in separation of high molecular weight DNAs. 1 * 5 g
Catalog # ICNA0219399305
Supplier:  MP Biomedicals
ETHIDIUM BROMIDE. Molecular Biology ReagentPurity: 98%Ideal for fluorometric detection of double stranded nucleic acids in gel elect rophoresis. Also acts as an RNA polymerase inhibitor, and in separation of high molecular weight DNAs. 1 * 5 g
Catalog # ICNA0219399305
Supplier:  MP Biomedicals
Frequently Bought Together