Order Entry
Hungary
ContactUsLinkComponent
SONOREX SUPER RK 52 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 150X140X100 MM 1,8 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEAT ING 30-80°C (86-176°F), ULTRASONIC PEAK OUTPUT 240 W, HF-OUTPUT: 60 WEFF. 1 * 1 items
Catalog # BAND164-GB
SONOREX SUPER RK 52 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 150X140X100 MM 1,8 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEAT ING 30-80°C (86-176°F), ULTRASONIC PEAK OUTPUT 240 W, HF-OUTPUT: 60 WEFF. 1 * 1 items
Catalog # BAND164-GB
Frequently Bought Together