The bottom sieves are made of stainless steel trapezoidal resp. round perforation sheets metal. Larger openings mean faster throughput, less time in the mill and less wear. Bottom sieves with round perforation are recommended for size reduction of b 1 * 1 items
Catalog # FRIT16.6756.10
Supplier: FRITSCH
Frequently Bought Together