Order Entry
Hungary
ContactUsLinkComponent
Suitable for fast and gentle pre-crushing and fine comminution of medium-hard, brittle materials up to a Mohs hardness of 6. Max. feed size: (depends on material) 20 mm Max. throughput quantity: (depends on material and sieve size) 80 l/h Final fine 1 * 1 items
Catalog # FRIT16.6050.00
Supplier:  FRITSCH
Suitable for fast and gentle pre-crushing and fine comminution of medium-hard, brittle materials up to a Mohs hardness of 6. Max. feed size: (depends on material) 20 mm Max. throughput quantity: (depends on material and sieve size) 80 l/h Final fine 1 * 1 items
Catalog # FRIT16.6050.00
Supplier:  FRITSCH
Frequently Bought Together