Order Entry
Hungary
Orders LinkContactUsLinkComponent
Squeegee mop and handles
Squeegee mop and handles
Squeegee mop, white, 560 mm
Catalog # PPSA3400715
Squeegee mop and handles
Catalog # PPSA3400715
Squeegee mop and handles
Squeegee mop and handles
Squeegee mop, white, 560 mm
Price
27 800 Ft
Log in for availability
1 * 1 DB
Product Details & Documents
Squeegee mops and handles for hygienically sensitive areas.
  • Mop: PP (body), TPE (lip)
  • Hygienic squeegee is suitable with cleanroom and telescopical handles
  • Handles are anodised aluminum tubes
Specifications
  • Description:
    Squeegee mop, white, 560 mm
  • Seq:
    01