Epiregulin (EREG) BioAssay™ ELISA Kit (Human) is a sandwich enzyme immunoassay for in vitro quantitative measurement of EREG in human serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. 1 * 96 Tests
Catalog # USBI024861
Supplier: US Biological
Frequently Bought Together