Order Entry
Hungary
ContactUsLinkComponent
THERMO-MAT 96 colourless, 1 x 5 mats Reusable and autoclavable silicone mat for 96 well plates. Temperature range from -20 C to +120 C. Effective and uniform sealing when using in thermocyclers with high pressures lid 1 * 5 items
  732-3208
undefined
THERMO-MAT 96 colourless, 1 x 5 mats Reusable and autoclavable silicone mat for 96 well plates. Temperature range from -20 C to +120 C. Effective and uniform sealing when using in thermocyclers with high pressures lid 1 * 5 items
  732-3208
Frequently Bought Together