Order Entry
Hungary
ContactUsLinkComponent
Microplate 96 well Fluotrac 600 High Binding black/µclear F-bottom without lid steril
Catalog # GREI655097UMC
Supplier:  Greiner Bio-One
Microplate 96 well Fluotrac 600 High Binding black/µclear F-bottom without lid steril
Catalog # GREI655097UMC
Supplier:  Greiner Bio-One