Order Entry
Hungary
ContactUsLinkComponent
C18P x 5µm particle size x 120Å pore size, 10mm length x 4.0mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2POL-05G04S
C18P x 5µm particle size x 120Å pore size, 10mm length x 4.0mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2POL-05G04S
Frequently Bought Together