Order Entry
Hungary
ContactUsLinkComponent
DEAMIDATED GLIADIN PEPTIDES IGG ELISA KIT DEAMIDATED GLIADIN PEPTIDES IGG ELISA KIT IS A SOLID-PHASE ENZYME IMMUNOASSAY FOR THE QUA NTITATIVE DETECTION OF IGG ANTIBODIES AGAINST DEAMIDATED GLIADIN PEPTIDES (DGP) 1 KIT 1 * 1 KIT
Catalog # ABNOKA3998
Supplier:  Abnova
DEAMIDATED GLIADIN PEPTIDES IGG ELISA KIT DEAMIDATED GLIADIN PEPTIDES IGG ELISA KIT IS A SOLID-PHASE ENZYME IMMUNOASSAY FOR THE QUA NTITATIVE DETECTION OF IGG ANTIBODIES AGAINST DEAMIDATED GLIADIN PEPTIDES (DGP) 1 KIT 1 * 1 KIT
Catalog # ABNOKA3998
Supplier:  Abnova
Frequently Bought Together