qPCR detection of Human Papillomavirus 18. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPV18
Supplier: PRIMERDESIGN
Frequently Bought Together