qPCR detection of Treponema pallidum. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viru ses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-T.PAL
Supplier: PRIMERDESIGN
Frequently Bought Together