Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of all Group 2 Coronavirus genomes. Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HCOV2
Gyártó:  PRIMERDESIGN
qPCR detection of all Group 2 Coronavirus genomes. Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HCOV2
Gyártó:  PRIMERDESIGN
Frequently Bought Together