Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of Human Rhinovirus Subtype 16. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard c urve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-HRV16
Supplier:  PRIMERDESIGN
qPCR detection of Human Rhinovirus Subtype 16. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard c urve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-HRV16
Supplier:  PRIMERDESIGN
Frequently Bought Together