Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of Mycobacterium leprae and Mycobacterium lepromatosis. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-LEPROSY
Supplier:  PRIMERDESIGN
qPCR detection of Mycobacterium leprae and Mycobacterium lepromatosis. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-LEPROSY
Supplier:  PRIMERDESIGN
Frequently Bought Together