Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of Staphylococcus epidermidis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA gen ome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-S.EPI
Supplier:  PRIMERDESIGN
qPCR detection of Staphylococcus epidermidis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA gen ome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-S.EPI
Supplier:  PRIMERDESIGN
Frequently Bought Together