Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of Mycoplasma felis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endog enous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-M.FELIS
Supplier:  PRIMERDESIGN
qPCR detection of Mycoplasma felis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endog enous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-M.FELIS
Supplier:  PRIMERDESIGN
Frequently Bought Together