Order Entry
Hungary
ContactUsLinkComponent
qPCR detection of Maize Dwarf Mosaic Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MDMV
Supplier:  PRIMERDESIGN
qPCR detection of Maize Dwarf Mosaic Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MDMV
Supplier:  PRIMERDESIGN
Frequently Bought Together