Cetrimide (Tetradecyltrimethylammonium bromide) 1119-97-7 1 * 500 g
MOLE14921960-500G
: Molekula
Frequently Bought Together
Methyl methanesulphonate 99%
: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
: VWR Chemicals
n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
2-Propanol, Technical Grade
: Thermo Fisher Scientific
Pyridine ≥99%
: Thermo Fisher Scientific
n-Octane ≥98%
: Thermo Fisher Scientific
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Xylene (mixture of isomers), extra pure
: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
: Thermo Fisher Scientific
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
: Honeywell Chemicals
Aniline ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
: VWR Chemicals