Order Entry
Hungary
ContactUsLinkComponent
HCV-NS5-G2 protein.The protein contains the HCV-NS5 genotype 2 immunodominant regions.Hepatitis C Virus NS5 protein is purified by p roprietarychromatographic techniques.The hepatitis C virus (HCV) NS5A gene product is aphosphorylated 56 to 1 * 1 mg
Catalog # BIRBORB97955-1
Supplier:  Biorbyt
HCV-NS5-G2 protein.The protein contains the HCV-NS5 genotype 2 immunodominant regions.Hepatitis C Virus NS5 protein is purified by p roprietarychromatographic techniques.The hepatitis C virus (HCV) NS5A gene product is aphosphorylated 56 to 1 * 1 mg
Catalog # BIRBORB97955-1
Supplier:  Biorbyt