Sandwich Elisa kit for Quantitative Detection for Mouse Clusterin, Clusterin apolipoprotein J is a 75-80 kDa disulfide-linked hetero dimeric protein associated with the clearance of cellular debris and apoptosis. In humans, clusterin is enco 1 * 96 Tests
Catalog # BIRBORB96806-96
Supplier: Biorbyt
Frequently Bought Together