Order Entry
Hungary
ContactUsLinkComponent
Sandwich Elisa kit for Quantitative Detection for Mouse Neprilysin or CD10, Neprilysin, also known as membrane metallo-endopeptidase , neutral endopeptidase NEP, CD10, and common acute lymphoblastic leukemia antigen CALLA, is a zinc-dependen 1 * 96 Tests
Catalog # BIRBORB96795-96
Supplier:  Biorbyt
Sandwich Elisa kit for Quantitative Detection for Mouse Neprilysin or CD10, Neprilysin, also known as membrane metallo-endopeptidase , neutral endopeptidase NEP, CD10, and common acute lymphoblastic leukemia antigen CALLA, is a zinc-dependen 1 * 96 Tests
Catalog # BIRBORB96795-96
Supplier:  Biorbyt