Sandwich Elisa kit for Quantitative Detection for Mouse P-Cadherin, P-Cadherin, also known as Cadherin-3CDH3, is a protein that in h umans is encoded by the CDH3 gene. This gene is a classical cadherin from the cadherin superfamily. This gen 1 * 96 Tests
Catalog # BIRBORB96779-96
Supplier: Biorbyt
Frequently Bought Together