Order Entry
Hungary
ContactUsLinkComponent
Hi solid sis kit, axial icp inert+sheath. for use with high tds samples. incl inert sturman-masters s/chamber, v-groove neb, sheath gas torch, argon saturation accy + all necessary tubing 1 * 1 items
Catalog # VARI9910141000
Supplier:  VARIAN
Hi solid sis kit, axial icp inert+sheath. for use with high tds samples. incl inert sturman-masters s/chamber, v-groove neb, sheath gas torch, argon saturation accy + all necessary tubing 1 * 1 items
Catalog # VARI9910141000
Supplier:  VARIAN
Frequently Bought Together