Order Entry
Hungary
ContactUsLinkComponent
C18G x 3µm particle size x 300Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03N03S
C18G x 3µm particle size x 300Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03N03S
Frequently Bought Together