Order Entry
Hungary
ContactUsLinkComponent
Biotix 300 µL Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: Fle xFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, serial number f 1 * 960 items
Catalog # 613-2821
Supplier:  BIOTIX
Biotix 300 µL Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: Fle xFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, serial number f 1 * 960 items
Catalog # 613-2821
Supplier:  BIOTIX
Frequently Bought Together