Order Entry
Hungary
ContactUsLinkComponent
IDEAL FOR EVERYDAY ANALYSIS OF NUCLEIC ACIDS BY GELE ELECTROPHORESIS OR BLOTTING (NORTHERN OR SOUTHERN) AND IS ALSO SUITABLE FOR PRO TEIN APPLICATIONS. NO BACKGROUND FLUORESCENCE HAS BEEN DETECTED BY ETHIDIUM BROMIDE ASSAY. 1 * 500 g
Catalog # ABNOD0021-2
Supplier:  Abnova
IDEAL FOR EVERYDAY ANALYSIS OF NUCLEIC ACIDS BY GELE ELECTROPHORESIS OR BLOTTING (NORTHERN OR SOUTHERN) AND IS ALSO SUITABLE FOR PRO TEIN APPLICATIONS. NO BACKGROUND FLUORESCENCE HAS BEEN DETECTED BY ETHIDIUM BROMIDE ASSAY. 1 * 500 g
Catalog # ABNOD0021-2
Supplier:  Abnova
Frequently Bought Together