Order Entry
Hungary
ContactUsLinkComponent
Neptune 1000 µl pipette tips. racked, 8 racks of 96 tips/pack, 4 packs/case. universal fit ideal for both single and multichannel p ipettors, graduation marks. . 1 * 768 items
Catalog # BITX2162.96
Supplier:  BIOTIX
Neptune 1000 µl pipette tips. racked, 8 racks of 96 tips/pack, 4 packs/case. universal fit ideal for both single and multichannel p ipettors, graduation marks. . 1 * 768 items
Catalog # BITX2162.96
Supplier:  BIOTIX
Frequently Bought Together