Order Entry
Hungary
ContactUsLinkComponent
kit, klk5 (human) elisa kit, klk5 (human) elisa kit is an immunoassay for quantitative detection of human klk5 in serum, plasma, bod y fluids, tissue lysates or cell culture supernatant. 1 * 1 KIT
Catalog # ABNOKA2352
Supplier:  Abnova
kit, klk5 (human) elisa kit, klk5 (human) elisa kit is an immunoassay for quantitative detection of human klk5 in serum, plasma, bod y fluids, tissue lysates or cell culture supernatant. 1 * 1 KIT
Catalog # ABNOKA2352
Supplier:  Abnova