Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ClO2- Chlorites 1000 mg/l in H2O for IC 1 * 500 mL
Catalog # ACSDH028.01O.L5
Supplier:
CPACHEM
Expand image
ClO2- Chlorites 1000 mg/l in H2O for IC 1 * 500 mL
Catalog # ACSDH028.01O.L5
Supplier:
CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals